Assay Details
Target Gene Details
Entrez Gene ID: | 646658 |
Gene Name: | synapse differentiation inducing 1 like |
Gene Aliases: |
CAPUCIN, DSPC1, IFITMD4, TMEM90A |
Location: |
Chr.14:74405893-74477080 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYNDIG1L | NM_001105579.1 | NP_001099049.1 | ||
XM_017021600.1 | XP_016877089.1 | |||
DB473943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832829 | Chr.14:74377797 - 74545100 on Build GRCh38 | Loss | LTBP2 SYNDIG1L ISCA2 NPC2 MIR4709 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |