Assay Details
Target Gene Details
Entrez Gene ID: | 7528 |
Gene Name: | YY1 transcription factor |
Gene Aliases: |
DELTA, INO80S, NF-E1, UCRBP, YIN-YANG-1 |
Location: |
Chr.14:100238765-100279034 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YY1 | NM_003403.4 | NP_003394.1 | ||
BC037308.1 | AAH37308.1 | |||
BC065366.1 | AAH65366.1 | |||
M76541.1 | AAA59926.1 | |||
M77698.1 | AAA59467.1 | |||
Z14077.1 | CAA78455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635496 | Chr.14:100169585 - 100290965 on Build GRCh38 | Gain | SLC25A29 MIR6764 YY1 |
esv2749105 | Chr.14:100239917 - 100262290 on Build GRCh38 | Deletion | YY1 |
nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss | SLC25A47 MIR345 SLC25A29 WARS MIR6764 YY1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |