Assay Details
Target Gene Details
Entrez Gene ID: | 100885778 |
Gene Name: | NALCN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:100708325-101059286 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NALCN-AS1 | NR_047687.1 | |||
AK025806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3633350 | Chr.13:100754199 - 100765002 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)