Assay Details
Target Gene Details
Entrez Gene ID: | 4008 |
Gene Name: | LIM domain 7 |
Gene Aliases: |
FBX20, FBXO20, LOMP |
Location: |
Chr.13:75620434-75859870 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMO7 | NM_001306080.1 | NP_001293009.1 | ||
NM_005358.5 | NP_005349.3 | |||
AF330045.1 | AAL37480.1 | |||
AK092052.1 | ||||
DA313248.1 | ||||
FJ711162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss | LMO7 LMO7-AS1 UCHL3 |
nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain | COMMD6 CTAGE11P TBC1D4 LMO7 LOC105370259 KLF12 LINC00381 LMO7-AS1 UCHL3 LINC01078 LINC00347 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |