Assay Details
Target Gene Details
Entrez Gene ID: | 8874 |
Gene Name: | Rho guanine nucleotide exchange factor 7 |
Gene Aliases: |
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB |
Location: |
Chr.13:111114619-111305734 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF7 | NM_001113511.2 | NP_001106983.1 | ||
NM_001113512.2 | NP_001106984.1 | |||
NM_001320852.1 | NP_001307781.1 | |||
NM_145735.3 | NP_663788.1 | |||
XM_006719956.3 | XP_006720019.1 | |||
XM_011521133.2 | XP_011519435.1 | |||
XM_017020814.1 | XP_016876303.1 | |||
XM_017020815.1 | XP_016876304.1 | |||
XM_017020817.1 | XP_016876306.1 | |||
AB177848.1 | BAD66826.1 | |||
AL834228.1 | CAD38906.1 | |||
BC033905.1 | AAH33905.1 | |||
BC060776.1 | AAH60776.1 |
Target Gene Details
Entrez Gene ID: | 100874226 |
Gene Name: | ARHGEF7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:111144305-111148565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF7-AS1 | NR_046516.1 | |||
DB508209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32604 | Chr.13:111140101 - 111157064 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2759967 | Chr.13:111009686 - 111154564 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)