Assay Details
Target Gene Details
Entrez Gene ID: | 10867 |
Gene Name: | tetraspanin 9 |
Gene Aliases: |
NET-5, NET5, PP1057 |
Location: |
Chr.12:3077355-3286564 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN9 | NM_001168320.1 | NP_001161792.1 | ||
NM_006675.4 | NP_006666.1 | |||
XM_011520912.2 | XP_011519214.1 | |||
AF089749.1 | AAC35859.1 | |||
AK027366.1 | ||||
AK223179.1 | BAD96899.1 | |||
BC071881.1 | AAH71881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832312 | Chr.12:3133005 - 3358034 on Build GRCh38 | Loss | LOC100128253 LOC105369606 TSPAN9 |
nsv557084 | Chr.12:3136424 - 3146196 on Build GRCh38 | Loss | TSPAN9 |
nsv527090 | Chr.12:3131129 - 3146196 on Build GRCh38 | Loss | TSPAN9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |