Assay Details
Target Gene Details
Entrez Gene ID: | 84876 |
Gene Name: | ORAI calcium release-activated calcium modulator 1 |
Gene Aliases: |
CRACM1, IMD9, ORAT1, TAM2, TMEM142A |
Location: |
Chr.12:121626550-121642040 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ORAI1 | NM_032790.3 | NP_116179.2 | ||
AK027372.1 | BAB55068.1 | |||
BC013386.1 | AAH13386.1 | |||
BC015369.1 | AAH15369.2 | |||
BC075831.1 | AAH75831.1 | |||
BC104633.1 | AAI04634.1 | |||
BG775018.1 | ||||
BM762473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560458 | Chr.12:121598620 - 121715590 on Build GRCh38 | Loss | TMEM120B MORN3 ORAI1 |
nsv1053272 | Chr.12:121507127 - 121694571 on Build GRCh38 | Gain | MORN3 KDM2B ORAI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |