Assay Details
Target Gene Details
Entrez Gene ID: | 341392 |
Gene Name: | acyl-CoA synthetase medium-chain family member 4 |
Gene Aliases: |
- |
Location: |
Chr.12:7304332-7328373 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACSM4 | NM_001080454.1 | NP_001073923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv448e212 | Chr.12:7280402 - 7344654 on Build GRCh38 | Gain | CD163L1 ACSM4 |
nsv832325 | Chr.12:7141408 - 7316916 on Build GRCh38 | Gain+Loss | CLSTN3 ACSM4 PEX5 |
dgv1349n100 | Chr.12:7274380 - 7336757 on Build GRCh38 | Gain | CD163L1 ACSM4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |