Assay Details
Target Gene Details
Entrez Gene ID: | 2305 |
Gene Name: | forkhead box M1 |
Gene Aliases: |
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT |
Location: |
Chr.12:2857681-2877155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXM1 | NM_001243088.1 | NP_001230017.1 | ||
NM_001243089.1 | NP_001230018.1 | |||
NM_021953.3 | NP_068772.2 | |||
NM_202002.2 | NP_973731.1 | |||
NM_202003.2 | NP_973732.1 | |||
XM_005253676.3 | XP_005253733.1 | |||
XM_011520930.2 | XP_011519232.1 | |||
XM_011520931.2 | XP_011519233.1 | |||
XM_011520932.1 | XP_011519234.1 | |||
XM_011520933.1 | XP_011519235.1 | |||
XM_011520934.2 | XP_011519236.1 | |||
XM_011520935.1 | XP_011519237.1 | |||
AK291206.1 | ||||
AK313845.1 | ||||
BC006192.2 | AAH06192.1 | |||
BC006529.2 | AAH06529.1 | |||
BQ229497.1 | ||||
DB137846.1 | ||||
U74612.1 | AAC51128.1 | |||
U74613.1 | AAC51129.1 | |||
U83113.1 | AAC63595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain | NRIP2 ITFG2 LOC100507424 TULP3 RHNO1 LOC101929469 FOXM1 FKBP4 |
dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss | LOC283440 NRIP2 ITFG2 LOC100507424 TULP3 RHNO1 LOC101929469 FOXM1 FKBP4 |
nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss | TEAD4 NRIP2 ITFG2 LOC100507424 TULP3 RHNO1 LOC101929469 FOXM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |