Assay Details
Target Gene Details
Entrez Gene ID: | 23067 |
Gene Name: | SET domain containing 1B |
Gene Aliases: |
KMT2G, Set1B |
Location: |
Chr.12:121803665-121832656 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SETD1B | NM_015048.1 | NP_055863.1 | ||
XM_005253858.4 | XP_005253915.1 | |||
XM_006719296.3 | XP_006719359.1 | |||
JF813787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160065 | Chr.12:121780583 - 121859371 on Build GRCh38 | Deletion | LINC01089 HPD RHOF SETD1B TMEM120B |
esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss | LINC01089 HPD RHOF SETD1B TMEM120B |
nsv911 | Chr.12:121796618 - 121841519 on Build GRCh38 | Deletion | LINC01089 HPD SETD1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |