Assay Details
Target Gene Details
Entrez Gene ID: | 219541 |
Gene Name: | mediator complex subunit 19 |
Gene Aliases: |
DT2P1G7, LCMR1, MED19AS |
Location: |
Chr.11:57703709-57712323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED19 | NM_001317078.1 | NP_001304007.1 | ||
NM_153450.2 | NP_703151.2 | |||
AY148462.1 | AAN16075.1 | |||
BC009723.1 | ||||
BC037223.1 | AAH37223.1 | |||
DB457116.1 | ||||
EB386171.1 | ||||
HE646678.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3579728 | Chr.11:57686071 - 57726599 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv508633 | Chr.11:57652112 - 57792573 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832164 | Chr.11:57640144 - 57811353 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)