Assay Details
Target Gene Details
Entrez Gene ID: | 8050 |
Gene Name: | pyruvate dehydrogenase complex component X |
Gene Aliases: |
DLDBP, E3BP, OPDX, PDX1, proX |
Location: |
Chr.11:34915829-34996128 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDHX | NM_001135024.1 | NP_001128496.1 | ||
NM_001166158.1 | NP_001159630.1 | |||
NM_003477.2 | NP_003468.2 | |||
XM_011520390.1 | XP_011518692.1 | |||
AF001437.1 | AAB66315.1 | |||
AK301384.1 | ||||
AK312463.1 | ||||
BC010389.1 | AAH10389.1 | |||
BI915296.1 | ||||
BX648910.1 | ||||
DA573433.1 | ||||
U82328.1 | AAC39661.1 | |||
Y13145.1 | CAA73606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759817 | Chr.11:34891768 - 34942181 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv34511 | Chr.11:34892577 - 34921289 on Build GRCh38 | Loss |
![]() ![]() |
nsv832119 | Chr.11:34793981 - 34987823 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)