Assay Details
Target Gene Details
Entrez Gene ID: | 26011 |
Gene Name: | teneurin transmembrane protein 4 |
Gene Aliases: |
Doc4, ETM5, ODZ4, TNM4, Ten-M4 |
Location: |
Chr.11:78653283-79441019 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TENM4 | NM_001098816.2 | NP_001092286.2 | ||
XM_011544933.2 | XP_011543235.1 | |||
XM_017017525.1 | XP_016873014.1 | |||
XM_017017526.1 | XP_016873015.1 | |||
AB037723.3 | BAA92540.3 | |||
AK127705.1 | ||||
BX537983.1 | CAD97943.1 | |||
BX640737.1 | CAE45850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483084 | Chr.11:78645595 - 78818126 on Build GRCh38 | Loss |
![]() |
esv33883 | Chr.11:78677719 - 78688834 on Build GRCh38 | Loss |
![]() |
esv34127 | Chr.11:78661992 - 79102008 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)