Assay Details
Target Gene Details
Entrez Gene ID: | 81532 |
Gene Name: | MOB kinase activator 2 |
Gene Aliases: |
HCCA2 |
Location: |
Chr.11:1469448-1486779 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOB2 | NM_001172223.2 | NP_001165694.1 | ||
NM_053005.5 | NP_443731.2 | |||
AB028173.1 | BAB18568.1 | |||
AB066564.1 | BAB62266.1 | |||
AJ580639.1 | CAE45271.1 | |||
AK057350.1 | BAB71443.1 | |||
AK296658.1 | ||||
AK300235.1 | ||||
BC047291.1 | AAH47291.1 | |||
BC067785.1 | AAH67785.1 | |||
DC413532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951279 | Chr.11:1337871 - 1491070 on Build GRCh38 | Deletion |
![]() ![]() |
dgv10n68 | Chr.11:1363751 - 1500210 on Build GRCh38 | Loss |
![]() ![]() |
esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)