Assay Details
Target Gene Details
Entrez Gene ID: | 161 |
Gene Name: | adaptor related protein complex 2 alpha 2 subunit |
Gene Aliases: |
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ |
Location: |
Chr.11:925809-1012245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A2 | NM_001242837.1 | NP_001229766.1 | ||
NM_012305.3 | NP_036437.1 | |||
XM_011519929.1 | XP_011518231.1 | |||
XM_011519930.2 | XP_011518232.1 | |||
AB020706.2 | BAA74922.2 | |||
AK001986.1 | ||||
AK027891.1 | BAB55435.1 | |||
AK223558.1 | BAD97278.1 | |||
AK293763.1 | ||||
AK299364.1 | ||||
BC006155.1 | AAH06155.1 | |||
BG705558.1 | ||||
BI462785.1 | ||||
DB461570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625088 | Chr.11:876861 - 979340 on Build GRCh38 | Gain | AP2A2 CHID1 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | MUC5AC DEAF1 CRACR2B PANO1 EPS8L2 CEND1 DRD4 MUC5B LMNTD2 PKP3 MIR6744 LOC101059906 CD151 RPLP2 LOC101927503 MIR210HG ANO9 LRRC56 TSPAN4 RASSF7 MIR210 TALDO1 RNH1 PNPLA2 TMEM80 PHRF1 MUC2 HRAS POLR2L LOC171391 LOC143666 SNORA52 B4GALNT4 CHID1 SIGIRR PIDD1 TOLLIP SLC25A22 AP2A2 CDHR5 PTDSS2 PDDC1 MUC6 SCT IRF7 |
dgv175e214 | Chr.11:851409 - 1043855 on Build GRCh38 | Gain | AP2A2 MUC6 TSPAN4 CHID1 |
nsv428244 | Chr.11:882364 - 1061595 on Build GRCh38 | Loss | AP2A2 MUC6 LOC101927503 CHID1 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | MUC5AC DEAF1 CRACR2B PANO1 EPS8L2 CEND1 DRD4 MUC5B LMNTD2 MIR6744 LOC101059906 CD151 RPLP2 TOLLIP-AS1 LOC101927503 MIR210HG ANO9 LRRC56 TSPAN4 RASSF7 MIR210 TALDO1 RNH1 PNPLA2 TMEM80 PHRF1 MUC2 HRAS POLR2L LOC171391 LOC143666 SNORA52 CHID1 SIGIRR PIDD1 TOLLIP SLC25A22 AP2A2 CDHR5 PTDSS2 PDDC1 MUC6 SCT IRF7 |
esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion | MUC5AC MUC2 BRSK2 CHID1 MUC5B MIR6744 MOB2 TOLLIP AP2A2 TOLLIP-AS1 MUC6 LOC101927503 TSPAN4 |
esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion | MUC5AC MUC2 BRSK2 CHID1 MUC5B MIR6744 MOB2 TOLLIP AP2A2 TOLLIP-AS1 MUC6 LOC101927503 TSPAN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |