Assay Details
Target Gene Details
Entrez Gene ID: | 57559 |
Gene Name: | STAM binding protein like 1 |
Gene Aliases: |
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2 |
Location: |
Chr.10:88879368-88923502 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STAMBPL1 | NM_020799.3 | NP_065850.1 | ||
XM_006717928.2 | XP_006717991.1 | |||
XM_006717930.3 | XP_006717993.1 | |||
XM_011539983.1 | XP_011538285.1 | |||
XM_011539985.1 | XP_011538287.1 | |||
XM_017016453.1 | XP_016871942.1 | |||
XM_017016454.1 | XP_016871943.1 | |||
XM_017016455.1 | XP_016871944.1 | |||
XM_017016456.1 | XP_016871945.1 | |||
XM_017016457.1 | XP_016871946.1 | |||
AB037794.1 | BAA92611.1 | |||
AK056086.1 | ||||
BC010846.1 | AAH10846.2 | |||
BX649096.1 | ||||
CD244526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain | LIPM ACTA2-AS1 ANKRD22 FAS ACTA2 STAMBPL1 |
esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain | ACTA2-AS1 FAS-AS1 FAS ACTA2 STAMBPL1 |
esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain | LIPM ACTA2-AS1 FAS-AS1 LIPN LIPK ANKRD22 FAS ACTA2 STAMBPL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |