Assay Details
Target Gene Details
Entrez Gene ID: | 36 |
Gene Name: | acyl-CoA dehydrogenase, short/branched chain |
Gene Aliases: |
2-MEBCAD, ACAD7, SBCAD |
Location: |
Chr.10:123008913-123058290 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACADSB | NM_001609.3 | NP_001600.1 | ||
XM_017016194.1 | XP_016871683.1 | |||
AK298638.1 | ||||
AK314241.1 | ||||
AL831821.2 | CAD38535.2 | |||
BC013756.1 | AAH13756.1 | |||
DA964130.1 | ||||
KU177879.1 | ||||
KU177880.1 | ||||
U12778.1 | AAA74424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion | PSTK IKZF5 ACADSB |
nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain | FAM24A LOC399815 PSTK IKZF5 CUZD1 ACADSB C10orf88 FAM24B FAM24B-CUZD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |