Assay Details
Target Gene Details
Entrez Gene ID: | 219736 |
Gene Name: | storkhead box 1 |
Gene Aliases: |
C10orf24 |
Location: |
Chr.10:68827537-68895942 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STOX1 | NM_001130159.2 | NP_001123631.1 | ||
NM_001130160.2 | NP_001123632.1 | |||
NM_001130161.2 | NP_001123633.1 | |||
NM_152709.4 | NP_689922.3 | |||
XM_011539454.2 | XP_011537756.1 | |||
AK124681.1 | BAC85925.1 | |||
AY842014.1 | AAW48524.1 | |||
AY842015.1 | AAW48525.1 | |||
AY842016.1 | AAW48526.1 | |||
AY842017.1 | AAW48527.1 | |||
BC140011.1 | ||||
DA118241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv437688 | Chr.10:68850453 - 68866528 on Build GRCh38 | Loss | STOX1 |
dgv151e199 | Chr.10:68852253 - 68864038 on Build GRCh38 | Deletion | STOX1 |
esv3578967 | Chr.10:68858922 - 68861077 on Build GRCh38 | Loss | STOX1 |
nsv1161974 | Chr.10:68851487 - 68866574 on Build GRCh38 | Duplication | STOX1 |
esv2762873 | Chr.10:68852888 - 68872027 on Build GRCh38 | Loss | STOX1 |
esv3623730 | Chr.10:68852320 - 68864024 on Build GRCh38 | Loss | STOX1 |
nsv1045343 | Chr.10:68853524 - 68866574 on Build GRCh38 | Loss | STOX1 |
esv25403 | Chr.10:68852829 - 68862542 on Build GRCh38 | Loss | STOX1 |
nsv8685 | Chr.10:68852817 - 68868800 on Build GRCh38 | Loss | STOX1 |
esv2759764 | Chr.10:68697962 - 68871556 on Build GRCh38 | Loss | MIR1254-1 SNORD98 TET1 CCAR1 STOX1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |