Assay Details
Target Gene Details
Entrez Gene ID: | 401494 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 4 |
Gene Aliases: |
PTPLAD2 |
Location: |
Chr.9:21003621-21031712 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HACD4 | NM_001010915.4 | NP_001010915.2 | ||
NM_001321883.1 | NP_001308812.1 | |||
NM_001321903.1 | NP_001308832.1 | |||
XM_011517876.2 | XP_011516178.1 | |||
XM_017014713.1 | XP_016870202.1 | |||
XM_017014714.1 | XP_016870203.1 | |||
BC114215.1 | AAI14216.1 | |||
BX538052.1 | CAD97990.1 | |||
BX648779.1 | ||||
DA333010.1 | ||||
HY118500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017312 | Chr.9:20892953 - 21122463 on Build GRCh38 | Gain | IFNB1 FOCAD HACD4 |
esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss | KLHL9 MIR31HG IFNA7 IFNA17 CDKN2B IFNA1 CDKN2A FOCAD-AS1 IFNA22P ERVFRD-3 IFNA21 MIR31 MTAP IFNA13 IFNA5 IFNA10 IFNA6 IFNB1 FOCAD IFNA16 HACD4 CDKN2B-AS1 IFNA14 MIR491 IFNA2 CDKN2A-AS1 IFNA8 IFNW1 IFNE IFNA4 |
nsv1161847 | Chr.9:21006309 - 21091241 on Build GRCh38 | Deletion | IFNB1 HACD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |