Assay Details
Target Gene Details
Entrez Gene ID: | 8395 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 beta |
Gene Aliases: |
MSS4, STM7 |
Location: |
Chr.9:68705217-69009176 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIP5K1B | NM_001278253.1 | NP_001265182.1 | ||
NM_003558.3 | NP_003549.1 | |||
XM_005252261.3 | XP_005252318.1 | |||
XM_006717300.2 | XP_006717363.1 | |||
XM_006717301.1 | XP_006717364.1 | |||
XM_017015188.1 | XP_016870677.1 | |||
XM_017015190.1 | XP_016870679.1 | |||
XM_017015191.1 | XP_016870680.1 | |||
AK292734.1 | ||||
AK295587.1 | ||||
CB988216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026962 | Chr.9:68335100 - 68742689 on Build GRCh38 | Gain | PGM5 TMEM252 PIP5K1B PGM5-AS1 LOC105376072 LINC01506 |
esv2422258 | Chr.9:68619929 - 69048424 on Build GRCh38 | Duplication | PRKACG FAM122A LOC101927069 PIP5K1B FXN LOC105376072 |
esv3302888 | Chr.9:68676237 - 68721189 on Build GRCh38 | Tandem Duplication | PIP5K1B |
esv3891677 | Chr.9:68431371 - 68730207 on Build GRCh38 | Gain | PGM5 TMEM252 PIP5K1B LOC105376072 LINC01506 |
esv3620546 | Chr.9:68675237 - 68722866 on Build GRCh38 | Gain | PIP5K1B |
esv2759693 | Chr.9:68676191 - 68738827 on Build GRCh38 | Gain | PIP5K1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |