Assay Details
Target Gene Details
Entrez Gene ID: | 29088 |
Gene Name: | mitochondrial ribosomal protein L15 |
Gene Aliases: |
HSPC145, L15mt, MRP-L15, MRP-L7, RPML7 |
Location: |
Chr.8:54135210-54148514 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL15 | NM_014175.3 | NP_054894.1 | ||
AF161494.1 | AAF29109.1 | |||
AK312832.1 | ||||
AL136665.1 | CAB66600.1 | |||
BC000891.1 | AAH00891.1 | |||
CR533531.1 | CAG38562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026869 | Chr.8:54077172 - 54176800 on Build GRCh38 | Gain | LYPLA1 MRPL15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |