Assay Details
Target Gene Details
Entrez Gene ID: | 5157 |
Gene Name: | platelet derived growth factor receptor like |
Gene Aliases: |
PDGRL, PRLTS |
Location: |
Chr.8:17576433-17643133 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFRL | NM_006207.2 | NP_006198.1 | ||
AK289450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2764066 | Chr.8:17530064 - 17677066 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
esv3616489 | Chr.8:17490020 - 17737549 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv831246 | Chr.8:17430363 - 17621085 on Build GRCh38 | Gain |
![]() ![]() |
nsv831247 | Chr.8:17541589 - 17701918 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3616490 | Chr.8:17490532 - 17587102 on Build GRCh38 | Loss |
![]() ![]() |
esv34867 | Chr.8:17488520 - 17725642 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2759600 | Chr.8:17433671 - 17784021 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv989082 | Chr.8:17567552 - 17577819 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)