Assay Details
Target Gene Details
Entrez Gene ID: | 101927245 |
Gene Name: | vegetative cell wall protein gp1-like |
Gene Aliases: |
- |
Location: |
Chr.8:102654313-102730620 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927245 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss | ATP6V1C1 LOC101927245 AZIN1 AZIN1-AS1 UBR5 ODF1 LOC107986873 KLF10 LOC401472 |
dgv7278n100 | Chr.8:102664742 - 102716969 on Build GRCh38 | Loss | LOC101927245 |
esv3618232 | Chr.8:102659191 - 102675397 on Build GRCh38 | Loss | LOC101927245 |