Assay Details
Target Gene Details
Entrez Gene ID: | 9228 |
Gene Name: | DLG associated protein 2 |
Gene Aliases: |
DAP2, SAPAP2 |
Location: |
Chr.8:1501366-1708476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP2 | NM_001277161.1 | NP_001264090.1 | ||
NM_004745.4 | NP_004736.2 | |||
AF119817.1 | AAF34344.1 | |||
AF119818.1 | AAF34345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469881 | Chr.8:1426471 - 1613783 on Build GRCh38 | Loss | DLGAP2 DLGAP2-AS1 |
nsv1024325 | Chr.8:651043 - 3203837 on Build GRCh38 | Gain | LOC401442 MIR7160 LOC105377777 DLGAP2 ERICH1 CLN8 CSMD1 LOC105377774 LOC101927752 ERICH1-AS1 DLGAP2-AS1 MYOM2 MIR3674 LOC101928058 LOC286083 LOC100131395 KBTBD11 LOC105377781 LOC105377792 MIR596 LOC101927815 LOC105377776 KBTBD11-OT1 ARHGEF10 |
esv3576504 | Chr.8:951237 - 1906751 on Build GRCh38 | Gain | DLGAP2 CLN8 MIR596 LOC101927752 ERICH1-AS1 LOC105377776 DLGAP2-AS1 MIR3674 LOC286083 ARHGEF10 |
nsv609509 | Chr.8:1352944 - 1641722 on Build GRCh38 | Gain | DLGAP2 LOC105377776 DLGAP2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |