Assay Details
Target Gene Details
Entrez Gene ID: | 54927 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 3 |
Gene Aliases: |
MINOS3, Mic19, PPP1R22 |
Location: |
Chr.7:132784862-133082158 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHCHD3 | NM_001317177.1 | NP_001304106.1 | ||
NM_017812.3 | NP_060282.1 | |||
NR_133671.1 | ||||
AK000427.1 | BAA91157.1 | |||
AK129533.1 | ||||
AK310236.1 | ||||
AL516679.3 | ||||
BC011596.2 | AAH11596.1 | |||
BC014839.1 | AAH14839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1032205 | Chr.7:132550902 - 133348828 on Build GRCh38 | Gain | EXOC4 CHCHD3 MIR6133 MIR3654 LOC100506937 FLJ40288 LOC105375512 PLXNA4 |
esv2759564 | Chr.7:132636931 - 132859723 on Build GRCh38 | Gain | CHCHD3 LOC100506937 FLJ40288 PLXNA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |