Assay Details
Target Gene Details
Entrez Gene ID: | 81628 |
Gene Name: | TSC22 domain family member 4 |
Gene Aliases: |
THG-1, THG1, TILZ2 |
Location: |
Chr.7:100466519-100479279 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D4 | NM_001303043.1 | NP_001289972.1 | ||
NM_030935.4 | NP_112197.1 | |||
NR_130118.1 | ||||
NR_130119.1 | ||||
AJ133115.1 | CAB43491.1 | |||
AK296580.1 | ||||
AK297702.1 | ||||
AK314824.1 | ||||
BC001486.2 | AAH01486.1 | |||
BC001966.1 | AAH01966.1 | |||
BC002972.1 | AAH02972.1 | |||
BC010406.2 | AAH10406.1 | |||
BC031622.1 | AAH31622.1 | |||
BX341171.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | STAG3L5P-PVRIG2P-PILRB UFSP1 TRIP6 FBXO24 TRIM56 GPC2 SLC12A9 MUC3A AZGP1 STAG3L5P MIR4658 MIR106B MUC12 SPDYE3 PVRIG2P PILRA COPS6 ZKSCAN1 MOGAT3 PILRB EPO CNPY4 ZASP TFR2 PVRIG EPHB4 SRRT MCM7 MIR25 AZGP1P1 SAP25 VGF POP7 MIR6840 MEPCE PCOLCE-AS1 ZCWPW1 PMS2P1 NYAP1 NAT16 MUC17 GIGYF1 LOC102724094 PLOD3 FIS1 CLDN15 PCOLCE MIR4653 C7orf61 ACTL6B ZNHIT1 PPP1R35 STAG3 GNB2 GATS ZSCAN21 AGFG2 LRCH4 MIR93 MOSPD3 ZAN ACHE AP4M1 GAL3ST4 AP1S1 MIR6875 C7orf43 ZNF3 TAF6 SERPINE1 LOC105375429 TSC22D4 LAMTOR4 MBLAC1 |
nsv527050 | Chr.7:100458543 - 100528352 on Build GRCh38 | Loss | NYAP1 C7orf61 TSC22D4 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | STAG3L5P-PVRIG2P-PILRB PMS2P1 NYAP1 STAG3L5P SPDYE3 PVRIG2P PILRA C7orf61 MIR6840 MEPCE PPP1R35 PILRB TSC22D4 ZCWPW1 |
nsv951385 | Chr.7:100467278 - 100496477 on Build GRCh38 | Deletion | NYAP1 TSC22D4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |