Assay Details
Target Gene Details
Entrez Gene ID: | 5764 |
Gene Name: | pleiotrophin |
Gene Aliases: |
HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1 |
Location: |
Chr.7:137227341-137343865 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTN | NM_001321386.1 | NP_001308315.1 | ||
NM_001321387.1 | NP_001308316.1 | |||
NM_002825.5 | NP_002816.1 | |||
XM_017012473.1 | XP_016867962.1 | |||
AK290488.1 | ||||
AK313424.1 | ||||
BC005916.1 | AAH05916.1 | |||
BI462078.1 | ||||
BT019692.1 | AAV38498.1 | |||
BX441769.2 | ||||
CN413530.1 | ||||
CR450338.1 | CAG29334.1 | |||
D90226.1 | BAA14261.1 | |||
M57399.1 | AAA35961.1 | |||
X52946.1 | CAA37121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv11620n54 | Chr.7:137198178 - 137250712 on Build GRCh38 | Loss | PTN |
esv33947 | Chr.7:137235266 - 138283104 on Build GRCh38 | Gain | AKR1D1 PTN CREB3L2 MIR4468 LOC100130880 LOC105375527 DGKI |
nsv831149 | Chr.7:137146918 - 137321721 on Build GRCh38 | Gain | PTN LOC349160 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |