Assay Details
Target Gene Details
Entrez Gene ID: | 81628 |
Gene Name: | TSC22 domain family member 4 |
Gene Aliases: |
THG-1, THG1, TILZ2 |
Location: |
Chr.7:100466519-100479279 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D4 | NM_001303043.1 | NP_001289972.1 | ||
NM_030935.4 | NP_112197.1 | |||
NR_130118.1 | ||||
NR_130119.1 | ||||
AJ133115.1 | CAB43491.1 | |||
AK296580.1 | ||||
AK297702.1 | ||||
AK314824.1 | ||||
BC001486.2 | AAH01486.1 | |||
BC001966.1 | AAH01966.1 | |||
BC002972.1 | AAH02972.1 | |||
BC010406.2 | AAH10406.1 | |||
BX341171.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | AP4M1 MUC17 MIR25 STAG3 PMS2P1 LOC102724094 PCOLCE-AS1 TAF6 GATS GPC2 VGF CNPY4 UFSP1 MCM7 AP1S1 MOGAT3 PCOLCE TRIM56 SLC12A9 PLOD3 ZKSCAN1 MIR106B SERPINE1 PVRIG2P STAG3L5P GAL3ST4 GNB2 ZNF3 MIR6875 ACTL6B ZNHIT1 LRCH4 MIR6840 ZASP MOSPD3 ZCWPW1 SRRT TSC22D4 LAMTOR4 AGFG2 TRIP6 ZSCAN21 FBXO24 TFR2 ZAN POP7 AZGP1 CLDN15 MIR93 SAP25 PILRA PILRB C7orf43 EPHB4 MIR4658 GIGYF1 NYAP1 COPS6 NAT16 C7orf61 MBLAC1 LOC105375429 MUC12 MIR4653 SPDYE3 MUC3A ACHE STAG3L5P-PVRIG2P-PILRB MEPCE FIS1 PVRIG PPP1R35 EPO AZGP1P1 |
nsv527050 | Chr.7:100458543 - 100528352 on Build GRCh38 | Loss | NYAP1 C7orf61 TSC22D4 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | SPDYE3 PMS2P1 PVRIG2P STAG3L5P PILRA STAG3L5P-PVRIG2P-PILRB PILRB MEPCE MIR6840 NYAP1 C7orf61 PPP1R35 ZCWPW1 TSC22D4 |
nsv951385 | Chr.7:100467278 - 100496477 on Build GRCh38 | Deletion | NYAP1 TSC22D4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |