Assay Details
Target Gene Details
Entrez Gene ID: | 22999 |
Gene Name: | regulating synaptic membrane exocytosis 1 |
Gene Aliases: |
CORD7, RAB3IP2, RIM, RIM1 |
Location: |
Chr.6:71886703-72403143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830687 | Chr.6:71791684 - 71969857 on Build GRCh38 | Gain | RIMS1 |
nsv603640 | Chr.6:71347604 - 72468161 on Build GRCh38 | Gain | MIR30A RIMS1 LINC00472 LINC01626 MIR30C2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |