Assay Details
Target Gene Details
Entrez Gene ID: | 389362 |
Gene Name: | proteasome assembly chaperone 4 |
Gene Aliases: |
C6orf86, PAC4, bA506K6.2 |
Location: |
Chr.6:3254325-3308224 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG4 | NM_001128591.1 | NP_001122063.1 | ||
NM_001128592.1 | NP_001122064.1 | |||
NM_001135750.1 | NP_001129222.1 | |||
XM_011514591.2 | XP_011512893.1 | |||
XM_011514593.2 | XP_011512895.1 | |||
XM_011514594.2 | XP_011512896.1 | |||
XM_011514595.2 | XP_011512897.1 | |||
AK095683.1 | ||||
AK125314.1 | ||||
AK126623.1 | ||||
BM738331.1 | ||||
CN429483.1 | ||||
DA093618.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |