Assay Details
Target Gene Details
Entrez Gene ID: | 23607 |
Gene Name: | CD2 associated protein |
Gene Aliases: |
CMS |
Location: |
Chr.6:47477746-47627263 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD2AP | NM_012120.2 | NP_036252.1 | ||
XM_005248976.1 | XP_005249033.1 | |||
XM_017010641.1 | XP_016866130.1 | |||
AF146277.1 | AAD34595.1 | |||
BC069444.1 | AAH69444.1 | |||
DA040834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5290 | Chr.6:47452135 - 47490906 on Build GRCh38 | Insertion | CD2AP |
nsv603012 | Chr.6:47464901 - 47647672 on Build GRCh38 | Loss | CD2AP |
nsv471637 | Chr.6:47339291 - 47493663 on Build GRCh38 | Gain+Loss | CD2AP |
nsv830655 | Chr.6:47407526 - 47595578 on Build GRCh38 | Gain | CD2AP |
nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain | LOC101926962 ADGRF1 CD2AP OPN5 PTCHD4 TNFRSF21 ADGRF4 ADGRF2 ADGRF5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |