Assay Details
Target Gene Details
Entrez Gene ID: | 84539 |
Gene Name: | melanin concentrating hormone receptor 2 |
Gene Aliases: |
GPR145, GPRv17, MCH-2R, MCH-R2, MCH2, MCH2R, MCHR-2, SLT |
Location: |
Chr.6:99919910-99994238 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCHR2 | NM_001040179.1 | NP_001035269.1 | ||
NM_032503.2 | NP_115892.2 | |||
AF347063.1 | AAK32193.1 | |||
AK123634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752086 | Chr.6:99809403 - 100227174 on Build GRCh38 | Gain |
![]() ![]() |
dgv6112n100 | Chr.6:99829964 - 100202622 on Build GRCh38 | Gain |
![]() ![]() |
nsv604259 | Chr.6:99855390 - 100194991 on Build GRCh38 | Gain |
![]() ![]() |
nsv1024904 | Chr.6:99839200 - 100082800 on Build GRCh38 | Loss |
![]() ![]() |
esv3576203 | Chr.6:99830480 - 100195802 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)