Assay Details
Target Gene Details
Entrez Gene ID: | 253314 |
Gene Name: | eukaryotic translation initiation factor 4E family member 1B |
Gene Aliases: |
- |
Location: |
Chr.5:176630613-176646644 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF4E1B | NM_001099408.1 | NP_001092878.1 | ||
XM_011534499.2 | XP_011532801.1 | |||
AK094270.1 | ||||
DA689940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv600346 | Chr.5:176597879 - 176669235 on Build GRCh38 | Loss | GPRIN1 EIF4E1B SNCB MIR4281 TSPAN17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |