Assay Details
Target Gene Details
Entrez Gene ID: | 3094 |
Gene Name: | histidine triad nucleotide binding protein 1 |
Gene Aliases: |
HINT, NMAN, PKCI-1, PRKCNH1 |
Location: |
Chr.5:131159283-131165348 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HINT1 | NM_005340.6 | NP_005331.1 | ||
NR_024610.2 | ||||
NR_024611.2 | ||||
NR_073488.1 | ||||
NR_134494.1 | ||||
NR_134495.1 | ||||
XM_017009410.1 | XP_016864899.1 | |||
AK026557.1 | BAB15500.1 | |||
BC001287.1 | AAH01287.1 | |||
BC007090.1 | AAH07090.1 | |||
BF967059.1 | ||||
BF970603.1 | ||||
BG393782.1 | ||||
BG527380.1 | ||||
BG708765.1 | ||||
BG942577.1 | ||||
CD639104.1 | ||||
CK002748.1 | ||||
CN290233.1 | ||||
CN290239.1 | ||||
CR457048.1 | CAG33329.1 | |||
DB125655.1 | ||||
DB289264.1 | ||||
U27143.1 | AAA82926.1 | |||
U51004.1 | AAC71077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575881 | Chr.5:130637365 - 131735693 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv5808n100 | Chr.5:130705748 - 131180675 on Build GRCh38 | Gain |
![]() ![]() |
esv2759379 | Chr.5:131103304 - 131256219 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)