Assay Details
Target Gene Details
Entrez Gene ID: | 2870 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
GPRK6 |
Location: |
Chr.5:177426496-177442849 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRK6 | NM_001004105.2 | NP_001004105.1 | ||
NM_001004106.2 | NP_001004106.1 | |||
NM_002082.3 | NP_002073.2 | |||
XM_006714859.2 | XP_006714922.1 | |||
XM_011534537.2 | XP_011532839.1 | |||
AF040751.1 | AAC09273.1 | |||
AF040752.1 | AAC09274.1 | |||
BC009277.2 | AAH09277.1 | |||
BC017272.1 | AAH17272.1 | |||
BI258860.1 | ||||
BU507609.1 | ||||
BX382886.2 | ||||
L16862.1 | AAA60175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss | SLC34A1 F12 DBN1 LMAN2 MXD3 PRR7 GRK6 PFN3 PRR7-AS1 RGS14 |
nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss | RAB24 PRELID1 SLC34A1 F12 LMAN2 MXD3 GRK6 NSD1 PFN3 RGS14 |
dgv10184n54 | Chr.5:177357511 - 177469411 on Build GRCh38 | Loss | SLC34A1 F12 DBN1 PRR7 GRK6 PFN3 PRR7-AS1 RGS14 |
nsv1161313 | Chr.5:177356552 - 177463415 on Build GRCh38 | Duplication | SLC34A1 F12 DBN1 PRR7 GRK6 PFN3 PRR7-AS1 RGS14 |
nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication | PRELID1 F12 DBN1 LMAN2 MXD3 RGS14 DDX41 SLC34A1 PRR7 PDLIM7 GRK6 FAM193B PFN3 PRR7-AS1 DOK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |