Assay Details
Target Gene Details
Entrez Gene ID: | 91977 |
Gene Name: | myozenin 3 |
Gene Aliases: |
CS-3, CS3, FRP3 |
Location: |
Chr.5:150660841-150679368 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYOZ3 | NM_001122853.2 | NP_001116325.1 | ||
NM_133371.4 | NP_588612.2 | |||
AF480443.1 | AAL79336.1 | |||
AJ300586.1 | CAC83076.1 | |||
AJ300587.1 | CAC83077.1 | |||
AK313873.1 | ||||
BC069380.1 | AAH69380.1 | |||
BC069553.1 | AAH69553.1 | |||
BC074870.2 | AAH74870.1 | |||
BC074871.2 | AAH74871.1 | |||
DB069077.1 | ||||
DB277496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1148e214 | Chr.5:150652572 - 150670289 on Build GRCh38 | Gain |
![]() ![]() |
esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv599960 | Chr.5:150636553 - 150685982 on Build GRCh38 | Loss |
![]() ![]() |
nsv428129 | Chr.5:150641519 - 150906180 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33898 | Chr.5:150657871 - 150766463 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)