Assay Details
Target Gene Details
Entrez Gene ID: | 8310 |
Gene Name: | acyl-CoA oxidase 3, pristanoyl |
Gene Aliases: |
- |
Location: |
Chr.4:8366263-8440725 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOX3 | NM_001101667.1 | NP_001095137.1 | ||
NM_003501.2 | NP_003492.2 | |||
XM_005248011.4 | XP_005248068.1 | |||
XM_005248013.3 | XP_005248070.1 | |||
XM_011513565.2 | XP_011511867.1 | |||
XM_011513566.1 | XP_011511868.1 | |||
AK225080.1 | ||||
AK313242.1 | ||||
BC017053.1 | AAH17053.1 | |||
Y11411.1 | CAA72214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4223 | Chr.4:8346346 - 8373558 on Build GRCh38 | Insertion | ACOX3 |
nsv593637 | Chr.4:8352102 - 8388919 on Build GRCh38 | Loss | ACOX3 |
nsv470013 | Chr.4:8314211 - 8375331 on Build GRCh38 | Gain | ACOX3 |
nsv526508 | Chr.4:8356196 - 8369827 on Build GRCh38 | Loss | ACOX3 |
nsv1002794 | Chr.4:8108356 - 8489207 on Build GRCh38 | Gain | SH3TC1 ACOX3 HTRA3 TRMT44 ABLIM2 |
nsv1009011 | Chr.4:8223942 - 8423104 on Build GRCh38 | Loss | SH3TC1 ACOX3 HTRA3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |