Assay Details
Target Gene Details
Entrez Gene ID: | 7884 |
Gene Name: | stem-loop binding protein |
Gene Aliases: |
HBP |
Location: |
Chr.4:1692731-1712741 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLBP | NM_001306074.1 | NP_001293003.1 | ||
NM_001306075.1 | NP_001293004.1 | |||
NM_006527.3 | NP_006518.1 | |||
DC383752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428434 | Chr.4:1623665 - 1813048 on Build GRCh38 | Gain | FGFR3 TMEM129 FAM53A SLBP LETM1 TACC3 |
nsv949756 | Chr.4:1710274 - 1817773 on Build GRCh38 | Deletion | FGFR3 TMEM129 SLBP LETM1 TACC3 |
dgv8901n54 | Chr.4:1644637 - 1864631 on Build GRCh38 | Loss | FGFR3 TMEM129 FAM53A SLBP LETM1 TACC3 |
nsv593316 | Chr.4:1526039 - 1768630 on Build GRCh38 | Gain | TMEM129 FAM53A SLBP TACC3 |
nsv508989 | Chr.4:1623823 - 1846590 on Build GRCh38 | Insertion | FGFR3 TMEM129 FAM53A SLBP LETM1 TACC3 |
nsv1161027 | Chr.4:1699833 - 1750693 on Build GRCh38 | Deletion | TMEM129 SLBP TACC3 |
nsv470000 | Chr.4:1522203 - 1722852 on Build GRCh38 | Loss | TMEM129 FAM53A SLBP TACC3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |