Assay Details
Target Gene Details
Entrez Gene ID: | 84803 |
Gene Name: | glycerol-3-phosphate acyltransferase 3 |
Gene Aliases: |
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1 |
Location: |
Chr.4:83535634-83605875 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPAT3 | NM_001256421.1 | NP_001243350.1 | ||
NM_001256422.1 | NP_001243351.1 | |||
NM_032717.4 | NP_116106.2 | |||
XM_011532384.2 | XP_011530686.1 | |||
XM_017008780.1 | XP_016864269.1 | |||
XM_017008781.1 | XP_016864270.1 | |||
AK055749.1 | BAB71002.1 | |||
AK223543.1 | BAD97263.1 | |||
AY358100.1 | AAQ88467.1 | |||
BC090956.1 | AAH90956.1 | |||
DA393801.1 | ||||
DA673378.1 | ||||
DQ324782.1 | ||||
DQ345298.1 | ABC70186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829994 | Chr.4:83445146 - 83637163 on Build GRCh38 | Loss | FAM175A MRPS18C HELQ GPAT3 |
nsv829995 | Chr.4:83500542 - 83557306 on Build GRCh38 | Loss | GPAT3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |