Assay Details
Target Gene Details
Entrez Gene ID: | 100507012 |
Gene Name: | BMPR1B antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:94743668-94757533 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMPR1B-AS1 | NR_121610.1 | |||
AI150588.1 |