Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAND2-AS1 | NR_003679.2 | |||
NR_136192.1 | 4 | 1393 | ||
NR_136193.1 | 4 | 2090 | ||
NR_136194.1 | 4 | 1560 | ||
NR_136195.1 | ||||
NR_136196.1 | ||||
NR_136197.1 | 4 | 1464 | ||
NR_136198.1 | 3 | 1281 | ||
NR_136199.1 | 5 | 1603 | ||
NR_136200.1 | 4 | 2093 | ||
NR_136201.1 | 4 | 2260 | ||
AB075501.1 | 4 | 2001 | BAE45751.1 | |
AK021601.1 | 1 | 1740 | BAB13853.1 | |
AK054605.1 | ||||
AK123668.1 | ||||
DA863153.1 | ||||
DB292439.1 | ||||
DQ831955.1 | 1 | 153 | ||
DQ831956.1 | 1 | 153 | ||
EF529495.1 | ||||
EF529496.1 | ||||
EF529497.1 | ||||
EF529498.1 | 4 | 528 | ||
EF529499.1 | ||||
EF529500.1 | ||||
EF529501.1 | ||||
EF529502.1 | ||||
EF529503.1 | ||||
EF529504.1 | ||||
EF529505.1 | ||||
EF529506.1 | ||||
EF529507.1 | ||||
EF529508.1 | ||||
EF529509.1 | ||||
EU000853.1 | 4 | 1306 | ||
EU000854.1 | 4 | 1306 | ||
EU000855.1 | 3 | 2398 | ||
EU000856.1 | 3 | 2398 |