Assay Details
Target Gene Details
Entrez Gene ID: | 51095 |
Gene Name: | tRNA nucleotidyl transferase 1 |
Gene Aliases: |
CCA1, CGI-47, MtCCA, RPEM, SIFD |
Location: |
Chr.3:3126916-3153435 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRNT1 | NM_001302946.1 | NP_001289875.1 | ||
NM_182916.2 | NP_886552.2 | |||
XM_011533776.2 | XP_011532078.1 | |||
XM_011533777.2 | XP_011532079.1 | |||
XM_011533778.2 | XP_011532080.1 | |||
AB063105.1 | BAB70662.1 | |||
AF151805.1 | AAD34042.1 | |||
AK290411.1 | ||||
AL834397.1 | CAD39059.1 | |||
DB032432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss | TRNT1 CNTN4-AS1 IL5RA LOC100130207 CRBN CNTN4 |
esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain | TRNT1 CNTN4-AS1 IL5RA CRBN CNTN4 |
esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication | TRNT1 CNTN4-AS1 IL5RA LOC100130207 CRBN CNTN4-AS2 CNTN4 |
nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss | TRNT1 CNTN4-AS1 IL5RA CRBN CNTN4 |
dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain | TRNT1 CNTN4-AS1 IL5RA LOC100130207 CRBN CNTN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |