Assay Details
Target Gene Details
Entrez Gene ID: | 54986 |
Gene Name: | unc-51 like kinase 4 |
Gene Aliases: |
FAM7C1, REC01035 |
Location: |
Chr.3:41246599-41963020 on Build GRCh38 |
Assay Gene Location: | Within Intron 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ULK4 | NM_001322501.1 | NP_001309430.1 | ||
NM_017886.3 | NP_060356.2 | |||
NR_136342.1 | ||||
XM_011533880.2 | XP_011532182.1 | |||
XM_017006711.1 | XP_016862200.1 | |||
AK093396.1 | ||||
AK304341.1 | ||||
AL133104.1 | CAB61411.1 | |||
BC040739.1 | AAH40739.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |