Assay Details
Target Gene Details
Entrez Gene ID: | 80145 |
Gene Name: | THO complex 7 |
Gene Aliases: |
NIF3L1BP1, fSAP24, hTREX30 |
Location: |
Chr.3:63833870-63864478 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THOC7 | NM_001285387.2 | NP_001272316.1 | ||
NM_001285404.1 | NP_001272333.1 | |||
NM_025075.3 | NP_079351.2 | |||
XM_006713339.3 | XP_006713402.1 | |||
AF104914.1 | ||||
AI391685.1 | ||||
AI784549.1 | ||||
AK027098.1 | BAB15656.1 | |||
AK225110.1 | ||||
BC065012.1 | AAH65012.1 | |||
BI914906.1 | ||||
CR457373.1 | CAG33654.1 | |||
HY023143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1008751 | Chr.3:63595517 - 63862172 on Build GRCh38 | Gain | C3orf49 LOC100130345 THOC7 THOC7-AS1 SYNPR SNTN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |