Assay Details
Target Gene Details
Entrez Gene ID: | 54986 |
Gene Name: | unc-51 like kinase 4 |
Gene Aliases: |
FAM7C1, REC01035 |
Location: |
Chr.3:41246599-41963020 on Build GRCh38 |
Assay Gene Location: | Within Intron 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ULK4 | NM_001322501.1 | NP_001309430.1 | ||
NM_017886.3 | NP_060356.2 | |||
NR_136342.1 | ||||
XM_011533880.2 | XP_011532182.1 | |||
XM_017006711.1 | XP_016862200.1 | |||
AK093396.1 | ||||
AK304341.1 | ||||
AL133104.1 | CAB61411.1 | |||
BC040739.1 | AAH40739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521572 | Chr.3:41286691 - 41318043 on Build GRCh38 | Loss |
![]() |
nsv1002077 | Chr.3:41269871 - 41303354 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)