Assay Details
Target Gene Details
Entrez Gene ID: | 5333 |
Gene Name: | phospholipase C delta 1 |
Gene Aliases: |
NDNC3, PLC-III |
Location: |
Chr.3:38007496-38029663 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCD1 | NM_001130964.1 | NP_001124436.1 | ||
NM_006225.3 | NP_006216.2 | |||
NR_024071.1 | ||||
XM_017006622.1 | XP_016862111.1 | |||
AB209862.1 | BAD93099.1 | |||
AK090774.1 | ||||
AK098690.1 | ||||
AK127813.1 | ||||
AK292324.1 | ||||
BC050382.1 | AAH50382.2 | |||
BX647927.1 | CAI46087.1 | |||
U09117.1 | AAA73567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759140 | Chr.3:37789992 - 38062733 on Build GRCh38 | Loss | ITGA9 ITGA9-AS1 PLCD1 CTDSPL VILL MIR26A1 DLEC1 |
nsv590101 | Chr.3:38000691 - 38065526 on Build GRCh38 | Loss | PLCD1 VILL DLEC1 |
nsv590094 | Chr.3:37941266 - 38021881 on Build GRCh38 | Loss | PLCD1 CTDSPL VILL MIR26A1 |
nsv522354 | Chr.3:37985093 - 38021881 on Build GRCh38 | Loss | PLCD1 VILL |
nsv428416 | Chr.3:37864474 - 38062733 on Build GRCh38 | Loss | PLCD1 CTDSPL VILL MIR26A1 DLEC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |