Assay Details
Target Gene Details
Entrez Gene ID: | 60678 |
Gene Name: | eukaryotic elongation factor, selenocysteine-tRNA specific |
Gene Aliases: |
EFSEC, SELB |
Location: |
Chr.3:128153456-128426193 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EEFSEC | NM_021937.4 | NP_068756.2 | ||
XM_005247696.2 | XP_005247753.1 | |||
XM_017006975.1 | XP_016862464.1 | |||
XM_017006976.1 | XP_016862465.1 | |||
XM_017006977.1 | XP_016862466.1 | |||
XM_017006980.1 | XP_016862469.1 | |||
AF268872.1 | AAG13375.1 | |||
BC007933.2 | AAH07933.1 | |||
BG420135.1 | ||||
DB454660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002685 | Chr.3:127819453 - 128455596 on Build GRCh38 | Gain | EEFSEC MGLL RUVBL1 KBTBD12 RUVBL1-AS1 SEC61A1 |
esv32985 | Chr.3:128124111 - 128192840 on Build GRCh38 | Loss | EEFSEC RUVBL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |