Assay Details
Target Gene Details
Entrez Gene ID: | 440944 |
Gene Name: | THUMPD3 antisense RNA 1 |
Gene Aliases: |
SETD5-AS1 |
Location: |
Chr.3:9385264-9397494 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THUMPD3-AS1 | NR_027007.3 | |||
NR_132780.1 | ||||
NR_132781.1 | ||||
AK093640.1 | ||||
AK128398.1 | 1 | 1340 | BAC87421.1 | |
AL110136.1 | ||||
AW237468.1 | ||||
BX953112.1 | ||||
HY142091.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv277n21 | Chr.3:9390066 - 9453435 on Build GRCh38 | Loss | SETD5 THUMPD3-AS1 |