Assay Details
Target Gene Details
Entrez Gene ID: | 6936 |
Gene Name: | GC-rich sequence DNA-binding factor 2 |
Gene Aliases: |
C2orf3, DNABF, GCF, TCF9 |
Location: |
Chr.2:75662655-75710986 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCFC2 | NM_001201334.1 | NP_001188263.1 | ||
NM_003203.4 | NP_003194.3 | |||
XM_005264520.3 | XP_005264577.1 | |||
XM_011533074.2 | XP_011531376.1 | |||
AB026911.1 | BAA85386.1 | |||
AK097564.1 | ||||
BC064559.1 | AAH64559.1 | |||
EF158468.1 | ||||
EF158469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv706e214 | Chr.2:75581910 - 75690050 on Build GRCh38 | Gain | MRPL19 GCFC2 |
dgv20n14 | Chr.2:75596036 - 75687909 on Build GRCh38 | Gain | MRPL19 GCFC2 |
esv2759063 | Chr.2:75462279 - 75785697 on Build GRCh38 | Gain+Loss | MRPL19 EVA1A LOC101927884 GCFC2 |
dgv152e55 | Chr.2:75594376 - 75686819 on Build GRCh38 | Gain | MRPL19 GCFC2 |
nsv514988 | Chr.2:75632582 - 75690230 on Build GRCh38 | Loss | MRPL19 GCFC2 |
dgv5n43 | Chr.2:75591363 - 75680225 on Build GRCh38 | Gain | MRPL19 GCFC2 |
nsv441762 | Chr.2:75594779 - 75687246 on Build GRCh38 | Gain | MRPL19 GCFC2 |
nsv819219 | Chr.2:75662623 - 75689964 on Build GRCh38 | Gain | GCFC2 |
nsv834263 | Chr.2:75617574 - 75780540 on Build GRCh38 | Gain | MRPL19 GCFC2 |
esv3303063 | Chr.2:75593915 - 75690043 on Build GRCh38 | Tandem Duplication | MRPL19 GCFC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |