Assay Details
Target Gene Details
Entrez Gene ID: | 8440 |
Gene Name: | NCK adaptor protein 2 |
Gene Aliases: |
GRB4, NCKbeta |
Location: |
Chr.2:105744649-105894274 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCK2 | NM_001004722.3 | NP_001004722.1 | ||
NM_003581.4 | NP_003572.2 | |||
XM_006712797.3 | XP_006712860.1 | |||
XM_011511991.2 | XP_011510293.1 | |||
XM_017005103.1 | XP_016860592.1 | |||
XM_017005105.1 | XP_016860594.1 | |||
BG480643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834321 | Chr.2:105663236 - 105841053 on Build GRCh38 | Loss |
![]() |
esv3591838 | Chr.2:105752843 - 105763169 on Build GRCh38 | Loss |
![]() |
esv3591832 | Chr.2:105718584 - 105763440 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)